accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4385-78-8

IUPAC Name: 3-(pyridin-4-yl)benzoate
Synonyms: 3-(pyridin-4-yl)benzoate
Molecular Weight (g/mol): 198.2
Molecular Formula: C12H8NO2
InChi Key: IYGIZNZSONLPSI-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=CC(=C1)C1=CC=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results