Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Pyrid-4-ylbenzoic acid, 97%, Thermo Scientific™

Catalog No. CC35601DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC35601DE 5 g
CC35601CB 250 mg
CC35601DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC35601DE Supplier Thermo Scientific Chemicals Supplier No. CC35601DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4385-78-8
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
InChI Key IYGIZNZSONLPSI-UHFFFAOYSA-N
Synonym 3-pyridin-4-yl benzoic acid, 3-pyridin-4-yl-benzoic acid, 3-pyrid-4-ylbenzoic acid, 3-4-pyridyl benzoic acid, 4-3-carboxyphenyl pyridine, 3-4-pyridinyl benzoic acid, 3-pyridin-4-yl benzoic acid, benzoic acid,3-4-pyridinyl, 5fl
PubChem CID 2795562
IUPAC Name 3-pyridin-4-ylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=CC=NC=C2

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 199.21
Percent Purity 97%
Chemical Name or Material 3-Pyrid-4-ylbenzoic acid
Hazard Category Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.