accessibility menu, dialog, popup

UserName

Viewing profile as user:
NONOOF

CAS RN 439097-58-2

IUPAC Name: 1-(2-fluoro-4-nitrophenyl)piperidin-4-one
Synonyms: 1-(2-fluoro-4-nitrophenyl)piperidin-4-one
Molecular Weight (g/mol): 238.22
Molecular Formula: C11H11FN2O3
InChi Key: BMXFDVAJYBTGRC-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(F)=C(C=C1)N1CCC(=O)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results