accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHNSO

CAS RN 439607-87-1

IUPAC Name: 5-bromo-2,3-dihydro-1,3-benzoxazole-2-thione
Synonyms: 5-bromo-3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol): 230.08
Molecular Formula: C7H4BrNOS
InChi Key: YFFBNUPHVZYFQL-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OC(=S)NC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results