accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NNO

CAS RN 439691-80-2

IUPAC Name: 4-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
Synonyms: 4-[(4-methylpiperazin-1-yl)methyl]benzaldehyde
Molecular Weight (g/mol): 218.3
Molecular Formula: C13H18N2O
InChi Key: DJJFKXUSAXIMLS-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=CC=C(C=O)C=C2)CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results