accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClSOONOO

CAS RN 4403-36-5

IUPAC Name: 2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)ethane-1-sulfonyl chloride
Synonyms: 2-(1,3-dioxoisoindol-2-yl)ethanesulfonyl chloride
Molecular Weight (g/mol): 273.69
Molecular Formula: C10H8ClNO4S
InChi Key: HCPVYBCAYPMANM-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)CCN1C(=O)C2=CC=CC=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results