Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Phthalimidoethanesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. P20495G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20495G 5 g
P20491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P20495G Supplier TCI America Supplier No. P20495G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4403-36-5
Molecular Formula C10H8ClNO4S
Molecular Weight (g/mol) 273.687
MDL Number MFCD01861217
InChI Key HCPVYBCAYPMANM-UHFFFAOYSA-N
Synonym 2-phthalimidoethanesulfonyl chloride, 2-1,3-dioxoisoindolin-2-yl ethanesulfonylchloride, 2-1,3-dioxoisoindolin-2-yl ethanesulfonyl chloride, 2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl ethanesulfonyl chloride, 2-phthalimidoethanesulfonylchloride, 2-phthalimido ethanesulfonyl chloride, 2-1,3-dioxoisoindol-2-yl ethanesulfonyl chloride, 2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl ethane-1-sulfonyl chloride, 2-1,3-dioxo-1,3-dihydro-isoindol-2-yl-ethanesulfonyl chloride, 2-2-chlorosulfonyl ethyl benzo c azoline-1,3-dione
PubChem CID 308739
IUPAC Name 2-(1,3-dioxoisoindol-2-yl)ethanesulfonyl chloride
SMILES C1=CC=C2C(=C1)C(=O)N(C2=O)CCS(=O)(=O)Cl

Specifications

Melting Point 162°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 273.69
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Phthalimidoethanesulfonyl Chloride
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.