CAS RN 4408-60-0
IUPAC Name:
2-(2,4,6-trimethylphenyl)acetic acid
Synonyms:
(2,4,6-trimethylphenyl)acetic acid
Molecular Weight (g/mol):
178.23
Molecular Formula:
C11H14O2
InChi Key:
CQWMQAKKAHTCSC-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(CC(O)=O)C(C)=C1