Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mesityleneacetic Acid 98.0+%, TCI America™
SDP

Catalog No. M12185G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12185G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12185G Supplier TCI America Supplier No. M12185G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4408-60-0
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00014368
InChI Key CQWMQAKKAHTCSC-UHFFFAOYSA-N
Synonym 2-mesitylacetic acid, mesitylacetic acid, 2,4,6-trimethylphenylacetic acid, 2,4,6-trimethylphenyl acetic acid, 2-2,4,6-trimethylphenyl acetic acid, mesityleneacetic acid, 2,4,6-trimethylbenzeneacetic acid, 2-mesityleneacetic acid, benzeneacetic acid, 2,4,6-trimethyl, 2,4,6-trimethyl-phenyl-acetic acid
PubChem CID 78123
IUPAC Name 2-(2,4,6-trimethylphenyl)acetic acid
SMILES CC1=CC(=C(C(=C1)C)CC(=O)O)C

Specifications

Melting Point 170°C
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Mesityleneacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.