accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4408-81-5

IUPAC Name: 2-{[(2R)-1-[bis(carboxylatomethyl)azaniumyl]propan-2-yl](carboxylatomethyl)azaniumyl}acetate
Synonyms: 2-{[(2R)-1-[bis(carboxylatomethyl)ammonio]propan-2-yl](carboxylatomethyl)ammonio}acetate
Molecular Weight (g/mol): 304.26
Molecular Formula: C11H16N2O8
InChi Key: XNCSCQSQSGDGES-SSDOTTSWSA-L
SMILES: C[C@H](C[NH+](CC([O-])=O)CC([O-])=O)[NH+](CC([O-])=O)CC([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results