Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Diaminopropane-N,N,N',N'-tetraacetic Acid 98.0+%, TCI America™
SDP

Catalog No. D1329500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1329500G 500 g
D132925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1329500G Supplier TCI America Supplier No. D1329500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4408-81-5 | C11H18N2O8 | 306.271 g/mol

Chemical Identifiers

CAS 4408-81-5
Molecular Formula C11H18N2O8
Molecular Weight (g/mol) 306.271
MDL Number MFCD00004286
InChI Key XNCSCQSQSGDGES-UHFFFAOYSA-N
Synonym pdta, 1,2-diaminopropane-n,n,n',n'-tetraacetic acid, medta, 1,2-diaminopropanetetraacetic acid, propylenediamine tetra-acetic acid, propanoldiaminetetracetic acid, acetic acid, propylenedinitrilo tetra, ccris 1250, 1,2-propylenediamine tetraacetic acid, propane-1,2-dinitrilotetraacetic acid
PubChem CID 20442
IUPAC Name 2-[2-[bis(carboxymethyl)amino]propyl-(carboxymethyl)amino]acetic acid
SMILES CC(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O

Specifications

Melting Point 237°C
Color White
Quantity 500 g
Formula Weight 306.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Diaminopropane-N,N,N′,N′-tetraacetic Acid
RTECSNumber AJ3500000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.