accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4409-11-4

IUPAC Name: 4-[(4-chlorophenyl)methyl]pyridine
Synonyms: 4-[(4-chlorophenyl)methyl]pyridine
Molecular Weight (g/mol): 203.67
Molecular Formula: C12H10ClN
InChi Key: OHKBVLWPESSWKC-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CC2=CC=NC=C2)C=C1
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)