Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Chlorobenzyl)pyridine, 97%, Thermo Scientific™

Catalog No. AAA1067336
Catalog No. AAA1067336 Supplier Thermo Scientific Chemicals Supplier No. A1067336

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4409-11-4
Molecular Formula C12H10ClN
MDL Number MFCD00006444
Beilstein 1367705
Synonym 4-4-chlorobenzyl pyridine, 4-4-chlorophenyl methyl pyridine, 4-4'-chlorobenzyl pyridine, 4-4-chloro-benzyl-pyridine, 4-para-chlorobenzyl-pyridine, pyridine, 4-4-chlorophenyl methyl, acmc-2097fd, 4-p-chlorobenzyl pyridine
InChI Key OHKBVLWPESSWKC-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1CC2=CC=NC=C2)Cl
IUPAC Name 4-[(4-chlorophenyl)methyl]pyridine
Molecular Weight (g/mol) 203.67
PubChem CID 78124
Formula Weight 203.67
Percent Purity 97%
Density 1.165
Refractive Index 1.59
Chemical Name or Material 4- (4-Chlorobenzyl)pyridine
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
EINECSNumber 224-560-2
RTECSNumber US7204100
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.