accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4411-26-1

IUPAC Name: 1-isothiocyanatoadamantane
Synonyms: 1-isothiocyanatoadamantane
Molecular Weight (g/mol): 193.31
Molecular Formula: C11H15NS
InChi Key: YPKFLUARLJRPQM-UHFFFAOYSA-N
SMILES: S=C=NC12CC3CC(CC(C3)C1)C2
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results