Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Adamantyl Isothiocyanate, TCI America™
SDP

Catalog No. I04855G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I04855G Supplier TCI America Supplier No. I04855G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4411-26-1
Molecular Formula C11H15NS
Molecular Weight (g/mol) 193.308
MDL Number MFCD00074736
InChI Key YPKFLUARLJRPQM-UHFFFAOYSA-N
Synonym 1-adamantyl isothiocyanate, isothiocyanic acid 1-adamantyl ester, 1-adamantylisothiocyanate, 1-isothiocyanato-adamantane, tricyclo 3.3.1.1'3,7 dec-1-yl isocyanate, tricyclo 3.3.1.13,7 decane, 1-isothiocyanato, adamantanisothiocyanate, 3r,5s,7s-1-isothiocyanatoadamantane, 1-isothiocyanatotricyclo 3.3.1.1∼3,7∼ decane, tricyclo 3.3.1.1 3,7 decane, 1-isothiocyanato
PubChem CID 107279
IUPAC Name 1-isothiocyanatoadamantane
SMILES C1C2CC3CC1CC(C2)(C3)N=C=S

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 5 g
Formula Weight 193.31
Physical Form Crystalline Powder
Chemical Name or Material 1-Adamantyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.