accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNN

CAS RN 4411-83-0

IUPAC Name: [2,2'-bipyridine]-6,6'-dicarbonitrile
Synonyms: [2,2'-bipyridine]-6,6'-dicarbonitrile
Molecular Weight (g/mol): 206.21
Molecular Formula: C12H6N4
InChi Key: KTYGOEZHIBSNMW-UHFFFAOYSA-N
SMILES: N#CC1=NC(=CC=C1)C1=CC=CC(=N1)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results