Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6,6'-Dicyano-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. D45411G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D45411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D45411G Supplier TCI America Supplier No. D45411G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity building block suitable for the synthesis of functional metal complexes.

Chemical Identifiers

CAS 4411-83-0
Molecular Formula C12H6N4
Molecular Weight (g/mol) 206.21
MDL Number MFCD00797251
InChI Key KTYGOEZHIBSNMW-UHFFFAOYSA-N
Synonym 2,2′C-Bipyridine-6,6′C-dicarbonitrile
PubChem CID 3491825
IUPAC Name [2,2'-bipyridine]-6,6'-dicarbonitrile
SMILES N#CC1=NC(=CC=C1)C1=CC=CC(=N1)C#N

Specifications

Melting Point 258°C
Color White-Yellow
Quantity 1 g
Formula Weight 206.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6,6′-Dicyano-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.