accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4415-88-7

IUPAC Name: 4,9-diazatricyclo[5.3.0.0²,⁶]decane-3,5,8,10-tetrone
Synonyms: 4,9-diazatricyclo[5.3.0.0²,⁶]decane-3,5,8,10-tetrone
Molecular Weight (g/mol): 194.15
Molecular Formula: C8H6N2O4
InChi Key: NWRWMFOKSXWCHV-UHFFFAOYSA-N
SMILES: O=C1NC(=O)C2C1C1C2C(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results