Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3,4-Cyclobutanetetracarboxdiimide 98.0+%, TCI America™
SDP

Catalog No. C2679200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2679200MG 200 mg
C26791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2679200MG Supplier TCI America Supplier No. C2679200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4415-88-7
Molecular Formula C8H6N2O4
Molecular Weight (g/mol) 194.146
InChI Key NWRWMFOKSXWCHV-UHFFFAOYSA-N
PubChem CID 338831
SMILES C12C(C3C1C(=O)NC3=O)C(=O)NC2=O

Specifications

Melting Point 350°C
Color Yellow
Quantity 200 mg
Formula Weight 194.15
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,3,4-Cyclobutanetetracarboxdiimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.