accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3COOCH3H3C

CAS RN 4421-10-7

IUPAC Name: 2,2-bis(propan-2-yl)-1,3-dioxolane
Synonyms: 2,2-diisopropyl-1,3-dioxolane
Molecular Weight (g/mol): 158.24
Molecular Formula: C9H18O2
InChi Key: BIYJJEWTEPSHGZ-UHFFFAOYSA-N
SMILES: CC(C)C1(OCCO1)C(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results