Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2-Diisopropyl-1,3-dioxolane 96.0+%, TCI America™
SDP

Catalog No. D112925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D112925G 25 g
D1129500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D112925G Supplier TCI America Supplier No. D112925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4421-10-7
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.24
MDL Number MFCD00020902
InChI Key BIYJJEWTEPSHGZ-UHFFFAOYSA-N
Synonym 2,2-diisopropyl-1,3-dioxolane, 1,3-dioxolane, 2,2-bis 1-methylethyl, diisopropyldioxolan, acmc-2097fh, biyjjewtepshgz-uhfffaoysa, diisopropyl ketone ethylene acetal, 2,2-bis methylethyl-1,3-dioxolane, 2,2-di propan-2-yl-1,3-dioxolane, 2,2-bis propan-2-yl-1,3-dioxolane
PubChem CID 78137
IUPAC Name 2,2-di(propan-2-yl)-1,3-dioxolane
SMILES CC(C)C1(OCCO1)C(C)C

Specifications

Color Colorless
Boiling Point 172°C
Quantity 25 g
UN Number 1993
Formula Weight 158.24
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2-Diisopropyl-1,3-dioxolane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.