accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4424-03-7

IUPAC Name: 3',6'-dihydroxyspiro[2,1λ⁶-benzoxathiole-3,9'-xanthene]-1,1-dione
Synonyms: 3',6'-dihydroxyspiro[2,1λ⁶-benzoxathiole-3,9'-xanthene]-1,1-dione
Molecular Weight (g/mol): 368.36
Molecular Formula: C19H12O6S
InChi Key: VJYGFSGAAOVTLC-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=CC(O)=CC=C3C22OS(=O)(=O)C3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results