accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4425-73-4

IUPAC Name: 2-(9H-fluoren-9-ylidene)acetic acid
Synonyms: fluoren-9-ylideneacetic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: TUWTVRGEVPWFIX-UHFFFAOYSA-N
SMILES: OC(=O)C=C1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)