Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9-Fluorenylideneacetic acid, 99%, Thermo Scientific™

Catalog No. AAA1923222
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1923222 100 g
AAA1923206 5 g
AAA1923214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1923222 Supplier Thermo Scientific Chemicals Supplier No. A1923222

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4425-73-4
Molecular Formula C15H10O2
Molecular Weight (g/mol) 222.243
MDL Number MFCD00040930
InChI Key TUWTVRGEVPWFIX-UHFFFAOYSA-N
Synonym 9-fluorenylideneacetic acid, 9h-fluoren-9-ylideneacetic acid, 2-9h-fluoren-9-ylidene acetic acid, acetic acid, 9h-fluoren-9-ylidene, fluoren-9-ylideneacetic acid, biphenylenecrylic acid, acmc-20alvm, 9-fluorenylidene acetic acid, fluoren-9-ylidene-acetic acid, acetic acid, 9-fluorenylidene
PubChem CID 96792
IUPAC Name 2-fluoren-9-ylideneacetic acid
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C2=CC(=O)O

Specifications

Melting Point 231°C to 233°C
Quantity 100 g
Formula Weight 222.24
Percent Purity 99%
Chemical Name or Material 9-Fluorenylideneacetic acid
EINECSNumber 224-604-0
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.