accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3FCl

CAS RN 443-83-4

IUPAC Name: 1-chloro-3-fluoro-2-methylbenzene
Synonyms: 1-chloro-3-fluoro-2-methylbenzene
Molecular Weight (g/mol): 144.57
Molecular Formula: C7H6ClF
InChi Key: FNPVYRJTBXHIPB-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results