Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-fluorotoluene 98.0+%, TCI America™
SDP

Catalog No. C118125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C118125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C118125G Supplier TCI America Supplier No. C118125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 443-83-4
Molecular Formula C7H6ClF
Molecular Weight (g/mol) 144.57
MDL Number MFCD00000570
InChI Key FNPVYRJTBXHIPB-UHFFFAOYSA-N
Synonym 2-chloro-6-fluorotoluene, benzene, 1-chloro-3-fluoro-2-methyl, 2-fluoro-6-chlorotoluene, 6-chloro-2-fluorotoluene, 1-chloro-3-fluoro-2-methyl-benzene, pubchem1607, acmc-209jxn, 2-chloro-6-fluoro toluene, 2-fluoro-6-chloro toluene, ksc236s4r
PubChem CID 9933
IUPAC Name 1-chloro-3-fluoro-2-methylbenzene
SMILES CC1=C(F)C=CC=C1Cl

Specifications

Color Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 1993
Formula Weight 144.57
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Chloro-6-fluorotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.