accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4432-77-3

IUPAC Name: tert-butyl(ethyl)amine
Synonyms: tert-butyl(ethyl)amine
Molecular Weight (g/mol): 101.19
Molecular Formula: C6H15N
InChi Key: XQOIBQBPAXOVGP-UHFFFAOYSA-N
SMILES: CCNC(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results