Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-tert-Butylethylamine 98.0+%, TCI America™
SDP

Catalog No. B08885ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B08885ML 5 mL
B088825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B08885ML Supplier TCI America Supplier No. B08885ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4432-77-3
Molecular Formula C6H15N
Molecular Weight (g/mol) 101.193
MDL Number MFCD00059367
InChI Key XQOIBQBPAXOVGP-UHFFFAOYSA-N
PubChem CID 521156
IUPAC Name N-ethyl-2-methylpropan-2-amine
SMILES CCNC(C)(C)C

Specifications

Color Yellow
Boiling Point 89°C
Quantity 5 mL
UN Number 2733
Formula Weight 101.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-tert-Butylethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.