accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4433-01-6

IUPAC Name: [2,2'-bipyridine]-3,3'-dicarboxylic acid
Synonyms: [2,2'-bipyridine]-3,3'-dicarboxylic acid
Molecular Weight (g/mol): 244.21
Molecular Formula: C12H8N2O4
InChi Key: KNVZVRWMLMPTTJ-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(N=CC=C1)C1=C(C=CC=N1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results