Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridine-3,3'-dicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. B36221G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36221G 1 g
B36225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36221G Supplier TCI America Supplier No. B36221G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 4433-01-6 | C12H8N2O4 | 244.21 g/mol

Chemical Identifiers

CAS 4433-01-6
Molecular Formula C12H8N2O4
Molecular Weight (g/mol) 244.21
MDL Number MFCD00226068
InChI Key KNVZVRWMLMPTTJ-UHFFFAOYSA-N
PubChem CID 681885
IUPAC Name 2-(3-carboxypyridin-2-yl)pyridine-3-carboxylic acid
SMILES C1=CC(=C(N=C1)C2=C(C=CC=N2)C(=O)O)C(=O)O

Specifications

Melting Point 261°C
Color White
Quantity 1 g
Synonym 2,2′C-Binicotinic Acid
Formula Weight 244.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Bipyridine-3,3′-dicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.