accessibility menu, dialog, popup

UserName

Viewing profile as user:
HONHOHNO

CAS RN 4433-40-3

IUPAC Name: 5-(hydroxymethyl)-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 5-hydroxymethyluracil|5HYDROXYMETHYLD2uracil|5-hydroxymethyl-D2-uracil
Molecular Weight (g/mol): 142.11
Molecular Formula: C5H6N2O3
InChi Key: JDBGXEHEIRGOBU-UHFFFAOYSA-N
SMILES: OCC1=CNC(=O)NC1=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results