accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSNHOOOHO

CAS RN 4434-61-1

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Synonyms: 2-{[(benzyloxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol): 283.34
Molecular Formula: C13H17NO4S
InChi Key: FPKHNNQXKZMOJJ-UHFFFAOYNA-N
SMILES: CSCCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results