Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Carbobenzoxy-DL-methionine 98.0+%, TCI America™
SDP

Catalog No. C06301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C06301G 1 g
C063010G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C06301G Supplier TCI America Supplier No. C06301G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4434-61-1
Molecular Formula C13H17NO4S
Molecular Weight (g/mol) 283.342
MDL Number MFCD00065123
InChI Key FPKHNNQXKZMOJJ-UHFFFAOYSA-N
Synonym N-Cbz-DL-methionine, Z-DL-Met-OH
PubChem CID 97806
IUPAC Name 4-methylsulfanyl-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CSCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 113°C
Color White
Quantity 1 g
Formula Weight 283.34
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material N-Carbobenzoxy-DL-methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.