accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OHHOHO

CAS RN 4435-50-1

IUPAC Name: butane-1,2,3-triol
Synonyms: 1,2,3-trihydroxybutane
Molecular Weight (g/mol): 106.12
Molecular Formula: C4H10O3
InChi Key: YAXKTBLXMTYWDQ-UHFFFAOYNA-N
SMILES: CC(O)C(O)CO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results