Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Butanetriol 90.0+%, TCI America™
SDP

Catalog No. B09185ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09185ML 5 mL
B091825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B09185ML Supplier TCI America Supplier No. B09185ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4435-50-1
Molecular Formula C4H10O3
Molecular Weight (g/mol) 106.121
MDL Number MFCD00059662
InChI Key YAXKTBLXMTYWDQ-UHFFFAOYSA-N
Synonym 1,2,3-Trihydroxybutane
PubChem CID 20497
IUPAC Name butane-1,2,3-triol
SMILES CC(C(CO)O)O

Specifications

Color Yellow
Boiling Point 175°C
Quantity 5 mL
Formula Weight 106.12
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Butanetriol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.