accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4435-53-4

IUPAC Name: 3-methoxybutyl acetate
Synonyms: 3-methoxybutyl acetate
Molecular Weight (g/mol): 146.19
Molecular Formula: C7H14O3
InChi Key: QMYGFTJCQFEDST-UHFFFAOYNA-N
SMILES: COC(C)CCOC(C)=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)