accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOH

CAS RN 4439-24-1

IUPAC Name: 2-(2-methylpropoxy)ethan-1-ol
Synonyms: ethanol, 2-(2-methylpropoxy)-
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: HHAPGMVKBLELOE-UHFFFAOYSA-N
SMILES: CC(C)COCCO
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results