accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2OO

CAS RN 4442-59-5

IUPAC Name: 1-(2,3-dihydro-1,4-benzodioxin-2-yl)methanamine
Synonyms: 1,4-benzodioxan-2-methylamine
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: JHNURUNMNRSGRO-UHFFFAOYNA-N
SMILES: NCC1COC2=CC=CC=C2O1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results