accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4445-06-1

IUPAC Name: octadecylcyclohexane
Synonyms: cyclohexane, octadecyl-
Molecular Weight (g/mol): 336.65
Molecular Formula: C24H48
InChi Key: MWWVNZNVTGBKQO-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC1CCCCC1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results