accessibility menu, dialog, popup

UserName

Viewing profile as user:
BFClHOOH

CAS RN 444666-39-1

IUPAC Name: (2-chloro-5-fluorophenyl)boronic acid
Synonyms: 2-chloro-5-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: OVTZJDSREVDCTJ-UHFFFAOYSA-N
SMILES: OB(O)C1=C(Cl)C=CC(F)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results