Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-5-fluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. C26665G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2666-5G 5 g
C26661G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2666-5G Supplier TCI America Supplier No. C26665G

missing translation for 'validMainframeMsg'

Specifications

CAS 444666-39-1
Melting Point 82°C
Color White
Molecular Formula C6H5BClFO2
Quantity 5 g
MDL Number MFCD06656272
Synonym 2-chloro-5-fluorobenzeneboronic acid, 2-chloro-5-fluorophenyl boronic acid, 2-chloro-5-fluorophenyl-boronic acid, 2-chloro-5-fluoro-phenyl boronic acid, boronic acid, 2-chloro-5-fluorophenyl
InChI Key OVTZJDSREVDCTJ-UHFFFAOYSA-N
SMILES B(C1=C(C=CC(=C1)F)Cl)(O)O
IUPAC Name (2-chloro-5-fluorophenyl)boronic acid
Molecular Weight (g/mol) 174.362
PubChem CID 22648116
Formula Weight 174.36
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.