CAS RN 444796-09-2
IUPAC Name:
2,7-dibromo-9-phenyl-9H-carbazole
Synonyms:
2,7-dibromo-9-phenylcarbazole
Molecular Weight (g/mol):
401.1
Molecular Formula:
C18H11Br2N
InChi Key:
MDXCDMSVFQIDGN-UHFFFAOYSA-N
SMILES:
BrC1=CC2=C(C=C1)C1=C(C=C(Br)C=C1)N2C1=CC=CC=C1
1
–
1
of
1
results
1
2,7-Dibromo-9-phenylcarbazole 98.0+%, TCI America™
CAS: 444796-09-2 Molecular Formula: C18H11Br2N Molecular Weight (g/mol): 401.101 InChI Key: MDXCDMSVFQIDGN-UHFFFAOYSA-N PubChem CID: 22392517 IUPAC Name: 2,7-dibromo-9-phenylcarbazole SMILES: C1=CC=C(C=C1)N2C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br