Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Dibromo-9-phenylcarbazole 98.0+%, TCI America™
SDP

Catalog No. D4832200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4832200MG 200 mg
D48321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4832200MG Supplier TCI America Supplier No. D4832200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 444796-09-2
Molecular Formula C18H11Br2N
Molecular Weight (g/mol) 401.101
InChI Key MDXCDMSVFQIDGN-UHFFFAOYSA-N
PubChem CID 22392517
IUPAC Name 2,7-dibromo-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=CC(=C3)Br)C4=C2C=C(C=C4)Br

Specifications

Melting Point 181°C
Color White
Quantity 200 mg
Formula Weight 401.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Dibromo-9-phenylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.