accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFBrCl

CAS RN 445-01-2

IUPAC Name: 4-bromo-1-chloro-2-(trifluoromethyl)benzene
Synonyms: 4-bromo-1-chloro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 259.45
Molecular Formula: C7H3BrClF3
InChi Key: XGOCKBMEZPNDPJ-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(Br)=CC=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)