Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-chlorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. C18465G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C18465G 5 g
C184625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C18465G Supplier TCI America Supplier No. C18465G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 445-01-2
Molecular Formula C7H3BrClF3
Molecular Weight (g/mol) 259.45
MDL Number MFCD00000601
InChI Key XGOCKBMEZPNDPJ-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzotrifluoride, 4-bromo-1-chloro-2-trifluoromethyl benzene, 2-chloro-5-bromobenzotrifluoride, 5-bromo-2-chloro-benzenetrifluoride, 5-bromo-2-chlorotrifluoride, 3-bromo-6-chlorobenzotrifluoride, benzene, 4-bromo-1-chloro-2-trifluoromethyl, 5-bromo-2-chloro-alpha,alpha,alpha-trifluorotoluene, 2-chloro-5-bromo-trifluorotoluene, 4-bromo-1-chloro-2-trifluoromethyl-benzene
PubChem CID 67959
IUPAC Name 4-bromo-1-chloro-2-(trifluoromethyl)benzene
SMILES FC(F)(F)C1=CC(Br)=CC=C1Cl

Specifications

Melting Point -21°C
Boiling Point 195°C
Quantity 5 g
Formula Weight 259.45
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-chlorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.