CAS RN 445430-58-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ IKK Inhibitor III, BMS-345541
IKK Inhibitor III, BMS-345541, CAS 445430-58-0, is a cell-permeable, potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC50 = ≈300 nM).