Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ IKK Inhibitor III, BMS-345541

Catalog No. 4014801MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-148-01MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-148-01MG Supplier MilliporeSigma™ Supplier No. 4014801MG
Add to Cart
Add to Cart

IKK Inhibitor III, BMS-345541, CAS 445430-58-0, is a cell-permeable, potent, selective, and allosteric site-binding inhibitor of IKK-2 (IC50 = ≈300 nM).

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Off-white
Formulation Supplied as bis-TFA salt.
Description 96% by HPLC
Target Proteins IKK-2
CAS 445430-58-0
Molecular Formula C14H17N5·2CF3CO2H
Formula Weight 483.4g/mol
Solubility DMSO
Content And Storage Hygroscopic, Protect from light
Form Solid
Quantity 1 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.