accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 4455-26-9

IUPAC Name: methyldioctylamine
Synonyms: methyldioctylamine
Molecular Weight (g/mol): 255.49
Molecular Formula: C17H37N
InChi Key: YJLYANLCNIKXMG-UHFFFAOYSA-N
SMILES: CCCCCCCCN(C)CCCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results