Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyldi-n-octylamine 97.0+%, TCI America™
SDP

Catalog No. M023225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M023225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M023225ML Supplier TCI America Supplier No. M023225ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4455-26-9
Molecular Formula C17H37N
Molecular Weight (g/mol) 255.49
MDL Number MFCD00009559
InChI Key YJLYANLCNIKXMG-UHFFFAOYSA-N
Synonym methyldioctylamine, n-methyldioctylamine, n-methyldi-n-octylamine, di octyl methylamine, n,n-dioctylmethylamine, 1-octanamine, n-methyl-n-octyl, di-n-octylmethylamine, n,n-dioctyl-n-methylamine, n-methyl-n-octyl-1-octanamine, dioctyl methyl amine
PubChem CID 78202
IUPAC Name N-methyl-N-octyloctan-1-amine
SMILES CCCCCCCCN(C)CCCCCCCC

Specifications

Color Yellow
Boiling Point 303°C
Quantity 25 mL
UN Number 2735
Formula Weight 255.49
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyldi-n-octylamine
EINECSNumber (2)-0176
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.