accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COBr

CAS RN 4457-67-4

IUPAC Name: 1-bromo-4-methoxybutane
Synonyms: 1-bromo-4-methoxybutane
Molecular Weight (g/mol): 167.05
Molecular Formula: C5H11BrO
InChi Key: ALOQTNHQNMYBDE-UHFFFAOYSA-N
SMILES: COCCCCBr
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results